Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sulfuric Acid, 4.0 Normal, Reagents
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Poly Bottle
Specifications
Specifications
pH | <1 |
Color | Colorless |
Boiling Point | 105°C |
Packaging | Poly Bottle |
Specific Gravity | 1.12 |
Quantity | 1 L |
Physical Form | Liquid |
Grade | Reagent |
Chemical Name or Material | Sulfuric Acid, 4.0 Normal |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction