Sulfuric Acid, 4.00N, Ricca

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 83221

Packaging Poly natural
Quantity 4L

 View more versions of this product

Catalog No. 8322-1

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    Chemical Name or Material Sulfuric Acid

    Specifications

    Quantity 4L
    Concentration or Composition (by Analyte or Components) 4.00N
    Packaging Poly natural
    Chemical Name or Material Sulfuric Acid

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit