Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sulfuric acid, 50% v/v aq. soln., Alfa Aesar™
Click to view available options
Quantity:
1L
Specifications
Quantity | 1L |
Formula Weight | 98.08 |
Physical Form | Liquid |
Chemical Name or Material | Sulfuric acid, 50% v/v aq. soln. |
Safety and Handling
P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H290-H314-H335
DOTInformation : Hazard Class: 8
EINECSNumber : 231-639-5
RTECSNumber : WS5600000
TSCA : Yes
Recommended Storage : Ambient temperatures