Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 5N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Cubetainer
Narrow Mouth Polyethylene Bottle

Chemical Identifiers

7732-18-5
H2SO4

High quality chemicals for laboratory and specialized industrial use

Quantity 4L
Formula Weight 98.08
Packaging Cubetainer
Specific Gravity 1L = 1.11kg
Chemical Name or Material Sulfuric Acid, 5N Volumetric Solution
Grade BAKER ANALYZED™ Reagent