Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 5N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Cubetainer
Narrow Mouth Polyethylene Bottle
7732-18-5
H2SO4
water,dihydrogen oxide,distilled water,purified water,water vapor,sterile water,water, purified,water, deionized,water, mineral,dihydrogen monoxide
7732-18-5
H2SO4
water,dihydrogen oxide,distilled water,purified water,water vapor,sterile water,water, purified,water, deionized,water, mineral,dihydrogen monoxide

High quality chemicals for laboratory and specialized industrial use

Quantity 1L
Formula Weight 98.08
Grade BAKER ANALYZED™ Reagent
Packaging Narrow Mouth Polyethylene Bottle
Specific Gravity 1L = 1.11kg
Chemical Name or Material Sulfuric Acid, 5N Volumetric Solution