Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 8 Normal, Unstandardized, Reagents

Catalog No. 01333706
Click to view available options
Quantity:
4 L
10 L
20 L

Sulfuric Acid, 8 Normal, Unstandardized, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sulfuric Acid, 8 Normal, Unstandardized
pH <1
Color Colorless
CAS 7732-18-5, 7664-93-9
Boiling Point 100°C
Packaging HDPE Winpak
Specific Gravity 1.2359
Quantity 20 L
Molecular Formula H2SO4/H2O
Physical Form Liquid
Synonym Sulphuric Acid, Dihydrogen Sulfate
Grade Reagent
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.