Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 90% (w/w) Aqueous Solution, Ricca Chemical

Manufacturer:  Ricca Chemical CompanySupplier Diversity Partner 8194.972.5

Packaging Cubitainer
Quantity 10L

 View more versions of this product

Catalog No. 81949725

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Chemical Identifiers

7664-93-9,7732-18-5
95.0,6.0
Sulfuric Acid
94.0,5.0
Liquid

Specifications

Clear colorless
7664-93-9,7732-18-5
95.0,6.0
90% (w/w) Aqueous Solution
Sulfuric Acid
10L
94.0,5.0
Liquid
Cubitainer

Safety and Handling

ShelfLife : 24 Months

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit