Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 95.0 to 98.0%, N.F. - F.C.C., J.T.Baker™

Click to view available options
Quantity:
500mL
2.5L
Packaging:
Glass Bottle
7664-93-9
H2O4S
98.072
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
CHEBI:26836
sulfuric acid
OS(=O)(=O)O
7664-93-9
H2O4S
98.072
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
CHEBI:26836
sulfuric acid
OS(=O)(=O)O
Quantity 2.5L
Formula Weight 98.08
Packaging Glass Bottle
Specific Gravity 1L = 1.84kg
Grade FCC/NF
Chemical Name or Material Sulfuric Acid
Assay 95.0 to 98.0%