Sulfuric acid, Acculute Standard Volumetric Solution, Final Concentration 2.0N, Alfa Aesar™

Manufacturer: Alfa Aesar 88568LS

Quantity 6 units

 View more versions of this product

Catalog No. AA88568LS

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 7664-93-9
    Physical Form Liquid
    Molecular Formula H2SO4
    Formula Weight 98.08
    MDL Number MFCD00064589
    Chemical Name or Material Sulfuric acid

    Specifications

    Formula Weight 98.08
    Quantity 6 units
    Color Colorless to pale yellow
    DOT Information DOT Class: 8, Packaging Group: II
    EINECS Number 231-639-5
    Physical Form Liquid
    Appearance Colorless to pale yellow liquid
    Molecular Formula H2SO4
    MDL Number MFCD00064589
    CAS 7664-93-9
    Concentration or Composition (by Analyte or Components) Sulfuric acid: 55%; Water: 45%
    Health Hazard 1 H314: Causes severe skin burns and eye damage.
    Health Hazard 2 In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Never add water to this product. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
    Odor Acrid
    Vapor Pressure 23 hPa (17mm Hg) at 20°C
    Chemical Name or Material Sulfuric acid

    In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Never add water to this product. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

    H314: Causes severe skin burns and eye damage.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit