Sulfuric Acid ACS AR, Macron Fine Chemicals™

macron

Manufacturer: Macron 287646

Packaging Glass Container
Quantity 2.5L

 View more versions of this product

Catalog No. 14-650-380

  null
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 7664-93-9
    Molecular Formula H2SO4
    Formula Weight 98.08
    Chemical Name or Material Sulfuric Acid
    Grade ACS Grade

    Specifications

    Quantity 2.5L
    Formula Weight 98.08
    CAS 7664-93-9
    Packaging Glass Container
    Molecular Formula H2SO4
    Chemical Name or Material Sulfuric Acid
    Grade ACS Grade

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit