Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric acid, for analysis, ca. 96% solution in water, ACROS Organics™

Click to view available options
Quantity:
500mL
1L
2.5L
10L
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
7664-93-9
H2O4S
98.08
QAOWNCQODCNURD-UHFFFAOYSA-N
oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil
1118
CHEBI:26836
sulfuric acid
OS(=O)(=O)O
7664-93-9
H2O4S
98.08
QAOWNCQODCNURD-UHFFFAOYSA-N
oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil
1118
CHEBI:26836
sulfuric acid
OS(=O)(=O)O
For analysis
Viscosity 21 mPa.s (25°C)
Residue after Ignition 0.0005% max
Chloride 0.5ppm max
Packaging Glass Bottle
Ignition Residue 0.0005% max.
Boiling Point 290.0°C
Density 1.8300g/mL
Color Colorless
Melting Point 10.0°C
Quantity 500mL
Specific Gravity 1.83
Linear Formula H2SO4
Fieser 01,470; 05,633; 06,558; 07,347; 09,441
Merck Index 15,9104
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol (generation of heat)
Formula Weight 98.08
Physical Form Liquid
Percent Purity 95 to 97%
Grade Analysis
Chemical Name or Material Sulfuric acid
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 231-639-5