Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
2.5L
Packaging:
Clear Glass Bottle
Clear PVC Coated Glass Bottle
Glass Bottle
Glass Bottle, Single ship
PVC Coated Glass Bottle
7664-93-9
H2O4S
98.072
QAOWNCQODCNURD-UHFFFAOYSA-N
oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil
1118
CHEBI:26836
sulfuric acid
OS(=O)(=O)O
7664-93-9
H2O4S
98.072
QAOWNCQODCNURD-UHFFFAOYSA-N
oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil
1118
CHEBI:26836
sulfuric acid
OS(=O)(=O)O

High quality chemicals for laboratory and specialized industrial use

Quantity 2.5L
Formula Weight 98.08
Grade BAKER ANALYZED™ ACS Reagent
Packaging Glass Bottle
Specific Gravity 1L = 1.84kg
Chemical Name or Material Sulfuric Acid