Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sulfuric Acid, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Clear Glass Bottle
Clear PVC Coated Glass Bottle
Glass Bottle
Glass Bottle, Single ship
PVC Coated Glass Bottle
Specifications
Packaging | Glass Bottle, Single ship |
Specific Gravity | 1L = 1.84kg |
Quantity | 500 mL |
Formula Weight | 98.08 |
Grade | BAKER ANALYZED™ ACS Reagent |
Chemical Name or Material | Sulfuric Acid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction