Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, BAKER INSTRA-ANALYZED™ Plus, J.T.Baker™

Click to view available options
Quantity:
500mL
2.5L
Packaging:
Poly Bottle

Chemical Identifiers

7664-93-9
H2O4S
98.072
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
CHEBI:26836

Acids for critical trace metal analysis; Quality is assured for up to 64 trace metals tested to very low parts-per-billion (ppb) levels

    For use in ICP (Inductively Coupled Plasma Spectrometry), Trace Metal Analysis; NOTE: Intended for nonfuel end purposes only
Quantity 500mL
SMILES OS(=O)(=O)O
IUPAC Name sulfuric acid
Formula Weight 98.08
Packaging Poly Bottle
Specific Gravity 1L = 1.84kg
Chemical Name or Material Sulfuric Acid
Grade BAKER INSTRA-ANALYZED™ Plus, Meets ACS reagent chemical requirements