Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric acid-d2, for NMR, 98 wt.% in D2O, 99.5+% atom D, ACROS Organics™

Click to view available options
Quantity:
5mL
50mL
100mL
Packaging:
Glass bottle
13813-19-9
H2O4S
100.084
MFCD00003542
QAOWNCQODCNURD-ZSJDYOACSA-N
sulfuric acid-d2,deuterosulfuric acid,dideuterosulfuric acid,sulphuric 2h2 acid,sulfuric acid-d2 solution,d2so4,dideuterosulfuric acid solution,2 h? sulfuric acid,sulfuric acid-d2, 98 wt% in deuterium oxide,deuterosulfuric acid min in d 2 o isotopic
160992
dideuterio sulfate
OS(=O)(=O)O
13813-19-9
H2O4S
100.084
MFCD00003542
QAOWNCQODCNURD-ZSJDYOACSA-N
sulfuric acid-d2,deuterosulfuric acid,dideuterosulfuric acid,sulphuric 2h2 acid,sulfuric acid-d2 solution,d2so4,dideuterosulfuric acid solution,2 h? sulfuric acid,sulfuric acid-d2, 98 wt% in deuterium oxide,deuterosulfuric acid min in d 2 o isotopic
160992
dideuterio sulfate
OS(=O)(=O)O
Color Colorless
Packaging Glass bottle
Density 1.8700g/mL
Quantity 50mL
Deuterium (D) 99.50% min.
Linear Formula D2SO4
Formula Weight 100.09
Physical Form Liquid
Percent Purity 97.5 to 102.5%
Grade NMR
Chemical Name or Material Sulfuric acid-d2, 99.5+ atom % D
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 237-464-0

TSCA : TSCA