Alfa Aesar™ Sulfuric acid, Environmental Grade Plus, 93-98%

Manufacturer: Alfa Aesar™ 38752AK

Quantity 250mL

 View more versions of this product

Catalog No. AA38752AK

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7664-93-9
    Formula Weight 98.08
    MDL Number MFCD00064589
    UN Number UN1830
    Merck Index 14,8974
    Chemical Name or Material Sulfuric acid
    Grade Environmental Grade Plus

    Specifications

    Physical Form Liquid, Double Sub-Boiling Quartz Distillation
    Formula Weight 98.08
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H314
    Causes severe skin burns and eye damage.
    TSCA TSCA
    MDL Number MFCD00064589
    CAS 7664-93-9
    UN Number UN1830
    Merck Index 14,8974
    Density 1.84g/mL
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: SULFURIC ACID
    Odor Acrid
    RTECS Number WS5600000
    Color Colorless to pale yellow
    Quantity 250mL
    Chemical Name or Material Sulfuric acid
    Assay 93 to 98%
    Grade Environmental Grade Plus

    GHS H Statement
    H314
    Causes severe skin burns and eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit