Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, FCC, 95-98%, Spectrum™

Click to view available options
Quantity:
500 mL
2.5 L
6 x 500 mL
4 x 2.5 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly-Coated Glass Bottle
7664-93-9
H2O4S
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
OS(O)(=O)=O
7664-93-9
H2O4S
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
OS(O)(=O)=O

S1736, 7664-93-9, H2SO4

Spectrum™ Sulfuric Acid, FCC is considered a general purpose food additive and is used to directly control pH during the processing of food by discouraging the growth of bacteria and spoilage microbes. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Quantity 6 x 500 mL
CAS Max % 1
Arsenic (As) 3 mg/kg
Percent Purity 95 to 98%
Iron (Fe) 0.0002
Lead (Pb) 5 mg/kg
Packaging Amber Glass Bottle
Selenium (Se) 2e-005