Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, ICP-OES, for Trace Metal Analysis, Thermo Scientific™

Click to view available options
Quantity:
500 mL
2.5L
7664-93-9,7732-18-5
H2O4S
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
CHEBI:26836
7664-93-9,7732-18-5
H2O4S
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
CHEBI:26836

Suitable for digestion of samples prior to trace elemental analysis via ICP-OES. Tested for 35 metals at ppb levels and also meets ACS testing specifications.

  • Suitable for trace metal analysis by ICP-OES
  • Tested for 35 metal impurities at ppb levels
  • Also meets general ACS testing specifications
  • Ideal for analysis of environmental samples
  • Available in 500 mL and 2.5 L poly coated glass bottles
Aluminum (Al) ≤ 30 ppb
Color 10 max. (APHA)
Quantity 500 mL
Ammonium ≤ 1 ppm
Appearance Clear, colorless liquid
Arsenic (As) ≤ 4 ppb
Barium (Ba) ≤ 10 ppb
Bismuth (Bi) ≤ 10 ppb
Boron (B) ≤ 10 ppb
Cadmium (Cd) ≤ 2 ppb
Calcium (Ca) ≤ 50 ppb
Cobalt (Co) ≤ 0.5 ppb
Chromium (Cr) ≤ 6 ppb
Chloride ≤ 0.1 ppm
Density 1.843
Copper (Cu) ≤ 1 ppb
Physical Form Liquid
Percent Purity 95.0 to 98.0%
Grade For trace metal analysis
Gold (Au) ≤ 10 ppb
Heavy Metals 500 ppb max.
Iron (Fe) ≤ 50 ppb
Lead (Pb) ≤ 0.5 ppb
Lithium (Li) ≤ 10 ppb
Manganese (Mn) ≤ 1 ppb
Magnesium (Mg) ≤ 7 ppb
Mercury (Hg) ≤ 5 ppb
Molybdenum (Mo) ≤ 10 ppb
Nickel (Ni) ≤ 2 ppb
Nitrate ≤ 0.2 ppm
Packaging Poly Coated Glass Bottle
Phosphate ≤ 0.5 ppm
Potassium (K) ≤ 500 ppb
Residue after Ignition ≤ 5 ppm
Selenium (Se) ≤ 50 ppb
Silver (Ag) ≤ 1 ppb
Sodium (Na) ≤ 500 ppb
Strontium (Sr) ≤ 5 ppb
Substances Reducing Permanganate ≤ 2 ppm
Tin (Sn) ≤ 5 ppb
Titanium (Ti) ≤ 10 ppb
Vanadium (V) ≤ 10 ppb
Zinc (Zn) ≤ 5 ppb
Zirconium (Zr) ≤ 10 ppb
Chemical Name or Material Sulfuric Acid
Show More Show Less