Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

Sulfuric acid, Macron Fine Chemicals™

macron

Manufacturer:  Avantor Performance Materials 287614

Quantity 500mL

 View more versions of this product

Catalog No. 14-650-378

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

7664-93-9
H2SO4
Viscous Liquid
Viscous Liquid
98.079
Sulfuric acid

Specifications

Colorless to slightly yellow
7664-93-9
H2SO4
Viscous Liquid
10°C
Sulfuric acid
500mL
Viscous Liquid
98.079
1.84g/cm3
337°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit