Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, Multi-Compendial, N.F., J.T.Baker™

Click to view available options
Quantity:
500mL
2.5L
Packaging:
Glass Bottle

Chemical Identifiers

7664-93-9
H2O4S
98.072
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
CHEBI:26836
sulfuric acid
OS(=O)(=O)O
Quantity 2.5L
Formula Weight 98.08
Packaging Glass Bottle
Specific Gravity 1L = 1.84kg
Chemical Name or Material Sulfuric Acid
Grade Multi-Compendial/NF