Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid Solution, Volumetric, for general laboratory use, 1N, Honeywell Burdick & Jackson

Click to view available options
Quantity:
6 x 1L
Packaging:
Plastic Bottle
7664-93-9,7732-18-5
H2O4S
98.072
QAOWNCQODCNURD-UHFFFAOYSA-N
oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil
1118
CHEBI:26836
sulfuric acid
OS(=O)(=O)O
7664-93-9,7732-18-5
H2O4S
98.072
QAOWNCQODCNURD-UHFFFAOYSA-N
oil of vitriol,sulphuric acid,dihydrogen sulfate,mattling acid,battery acid,dipping acid,acide sulfurique,electrolyte acid,acidum sulfuricum,vitriol brown oil
1118
CHEBI:26836
sulfuric acid
OS(=O)(=O)O
Quantity 6 x 1L
CAS Min % >95.0000%
CAS Max % <5.0000%
UN Number UN3264
Physical Form Buffer/Volumetric Solutions
Packaging Plastic Bottle
Chemical Name or Material Sulfuric Acid Solution

ShelfLife : 810 days from date of manufacture

DOTInformation : Transport Hazard Class: 8; Packing Group:3: III; Proper Shipping Name: Corrosive liquid, acidic, inorganic, n.o.s.