Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sulfuric Acid, 10% (w/w), Ricca Chemical
Click to view available options
Quantity:
1 L
2.5 L
10 L
20 L
55 gal.
Packaging:
Cubitainer
Natural Poly Bottle
Poly Drum
Safety-coated Glass Bottle
Specifications
Color | Colorless |
Concentration | 10% (w/w) |
CAS Min % | 89.0, 9.0 |
CAS Max % | 91.0, 11.0 |
Packaging | Poly Drum |
Solvent or Matrix | Aqueous Solution |
Quantity | 55 gal. |
Physical Form | Liquid |
Chemical Name or Material | Sulfuric Acid Solution |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction