Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, Technical, 66° Baume', Reagents

Catalog No. 01336803
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
20 L
190 L
Catalog No. Quantity
01-336-803 1 L
01-336-804 190 L
01-336-802 2.5 L
01-336-801 20 L
01-336-800 4 L
01-336-799 500 mL
6 options

Catalog No. 01-336-803

Supplier: Reagents Holdings Llc C51027001A

Only null left
Add to Cart
Add to Cart

Sulfuric Acid, Technical, 66° Baume', Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym Oil of Vitriol, Sulphuric Acid, Dihydrogen Sulfate
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH 0
Color Colorless
Boiling Point 260°C
Packaging Poly Bottle
Specific Gravity 1.8309
Quantity 1 L
Physical Form Liquid
Grade Technical
Chemical Name or Material Sulfuric Acid, Technical, 66° Baume'
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.