Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, Technical, 66° Baume', Reagents

Catalog No. p-200021159
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
20 L
190 L

Sulfuric Acid, Technical, 66° Baume', Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7664-93-9, 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym Oil of Vitriol, Sulphuric Acid, Dihydrogen Sulfate
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH 0
Color Colorless
Boiling Point 260°C
Packaging Poly Drum
Specific Gravity 1.8309
Quantity 190 L
Physical Form Liquid
Grade Technical
Chemical Name or Material Sulfuric Acid, Technical, 66° Baume'
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.