Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, Textile Grade 70% w/w, Reagents

Catalog No. 01336307
Click to view available options
Quantity:
1 L
4 L
15 gal.
55 gal.

Sulfuric Acid, Textile Grade 70% w/w, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7664-93-9, 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym Sulphuric Acid, Dihydrogen Sulfate
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH 0
Color Colorless to Tan
Boiling Point 165°C
Packaging Poly Drum
Specific Gravity 1.6134
Quantity 55 gal.
Physical Form Liquid
Percent Purity 70%
Grade Reagent
Chemical Name or Material Sulfuric Acid, Textile Grade 70% w/w
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.