Learn More
Sulfurous acid, ACS, 6.0% SO{2} min, Thermo Scientific Chemicals
Description
Sulfurous acid is used as a reducing agent, bleaching agent and disinfectant. It is utilized for bleaching of moistened wool, straw for hats, and other materials injured by chlorine. It finds application as a disinfectant as well as used externally for its parasiticidal effect in various skin diseases. Its derivative viz. sulfites are included in antiseptics, antifermentatives, and antizymotics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Appearance | Colourless liquid |
Color | Colorless |
Density | 1.03 g/cm3 at 20°C |
Odor | Pungent |
Sensitivity | Air Sensitive |
Quantity | 500 mL |
Physical Form | Liquid |
Solubility Information | Miscible with water. |
Formula Weight | 82.08 |
Grade | ACS |
Show More |
Safety and Handling
Hazard Category | H319-H335 |
Precautionary Statement | P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c |
DOTInformation | DOT Class: 8, Packaging Group: II |
EINECSNumber | 231-973-1 |
RTECSNumber | WT2775000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.