Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SULT1A1 Mouse anti-Human, Clone: OTI2C5, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525297

Catalog No. PIMA525297


Add to cart

Description

Description

The soluble sulfotransferases contribute to the elimination of xenobiotics, the activation of procarcinogens and the regulation of hormones by catalyzing the sulfate conjugation of these substances. Members of the three groups comprising this superfamily (namely SULT1, SULT2 and SULT3) show selectivity to certain substrate compounds. SULT1A1 (sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1), also known as STP or STP1, is a 295 amino acid protein that localizes to the cytoplasm and belongs to the sulfotransferase family. Functioning as a homodimer that is expressed in brain, liver, skin and lung tissue, SULT1A1 catalyzes the sulfate conjugation of catecholamines, phenolic drugs and neurotransmitters and, via this catalytic activity, plays a role in the elimination of a variety of compounds from the body. Additionally, SULT1A1 may be involved in the activation of carcinogenic N-hydroxyarylamines, indicating a possible role in carcinogenesis.
Specifications

Specifications

SULT1A1
Monoclonal
0.57 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P50225
Aryl sulfotransferase 1; HAST1/HAST2; Human aryl sulfotransferase mRNA complete cds; MGC131921; MGC5163; OTTHUMP00000162568; OTTHUMP00000162569; P PST 1; P PST; P-PST 1; Phenol sulfating phenol sulfotransferase 1; Phenol sulfotransferase 1; Phenol-sulfating phenol sulfotransferase 1; PST; ST1A1; ST1A1; ST1A3; STP; STP1; Sulfotransferase 1A1; Sulfotransferase family 1A phenol preferring member 1; Sulfotransferase family cytosolic 1A phenol preferring member 1; Sulfotransferase phenol prefer
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6817
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2C5
Unconjugated
SULT1A1
Liquid
SULT1A1
Full length human recombit protein of SULT1A1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit