Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SULT1A3 Mouse anti-Human, Clone: OTI1E8, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527401

Catalog No. PIMA527401


Add to cart

Description

Description

Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. This gene encodes a phenol sulfotransferase with thermolabile enzyme activity. Four sulfotransferase genes are located on the p arm of chromosome 16; this gene and SULT1A4 arose from a segmental duplication. This gene is the most centromeric of the four sulfotransferase genes. Exons of this gene overlap with exons of a gene that encodes a protein containing GIY-YIG domains (GIYD1). Multiple alternatively spliced variants that encode the same protein have been described.
Specifications

Specifications

SULT1A3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P0DMM9
HAST, HAST3, M-PST, ST1A3/ST1A4, ST1A5, STM, TL-PST, aryl sulfotransferase 1A3/1A4, catecholamine-sulfating phenol sulfotransferase, dopamine-specific sulfotransferase, monoamine-sulfating phenosulfotransferase, phenol sulfotransferase 1A5, placental estrogen sulfotransferase, sulfokinase, sulfotransferase 1A3/1A4
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6818
Immunohistochemistry (Paraffin), Western Blot
OTI1E8
Unconjugated
SULT1A3
Liquid
SULT1A3
Full length human recombit protein of SULT1A3 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit