Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SULT2B1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550797

Catalog No. PA550797


Add to cart

Description

Description

The antibody detects endogenous levels of total SULT2B1 protein.

Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure is similar among family members. This gene sulfates dehydroepiandrosterone but not 4-nitrophenol, a typical substrate for the phenol and estrogen sulfotransferase subfamilies. Two alternatively spliced variants that encode different isoforms have been described.
Specifications

Specifications

SULT2B1
Polyclonal
Unconjugated
SULT2B1
Liquid
SULT2B1
Fusion protein corresponding to residues near the C terminal of human sulfotransferase family, cytosolic, 2B, member 1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1.5 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
O00204
HSST2
Rabbit
IgG
100 μL
-20°C
Primary
6820
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit