Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sumatriptan Succinate, EP, 97.5-102%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
103628-48-4
C18H27N3O6S
413.49
PORMUFZNYQJOEI-UHFFFAOYSA-N
1-{3-[2-(dimethylamino)ethyl]-1H-indol-5-yl}-N-methylmethanesulfonamide; butanedioic acid
OC(=O)CCC(O)=O.CNS(=O)(=O)CC1=CC=C2NC=C(CCN(C)C)C2=C1
103628-48-4
C18H27N3O6S
413.49
PORMUFZNYQJOEI-UHFFFAOYSA-N
1-{3-[2-(dimethylamino)ethyl]-1H-indol-5-yl}-N-methylmethanesulfonamide; butanedioic acid
OC(=O)CCC(O)=O.CNS(=O)(=O)CC1=CC=C2NC=C(CCN(C)C)C2=C1

S3435, 103628-48-4, C18H27N3O6S

Spectrum™ Sumatriptan Succinate, EP , a synthetic drug part of the triptan class, is used to treat migraine headaches. The EP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Water 0.01
Appearance White
Quantity 100 g
pH 4.5 to 5.3
CAS Max % 1
Percent Purity 97.5 to 102%
Grade EP