Learn More
SUMO-tag Mouse, Clone: 4G11E9, GenScript™
Mouse Monoclonal Antibody
Supplier: Genscript Corporation A0169340

Description
Reconstitute the lyophilized powder with deionized water (or equivalent) to an antibody concentration of 0.5 mg/mL. SUMO tags are emerging as a viable alternative for increasing both the expression and solubility of otherwise hard-to-express proteins. The SUMO tag can be cleanly excised using SUMO protease, which recognizes the conformation of SUMO protein rather than a specific sequence within SUMO. The SUMO system can be used effectively in both prokaryotic and eukaryotic systems. GenScript SUMO-tag Antibody (4G11E9), mAb, Mouse is suitable for detecting fusion proteins that contain a SUMO-tag.GenScript SUMO-tag Antibody (4G11E9), mAb, Mouse is produced from the hybridoma resulting from fusion of Sp2/0 myeloma and lymphocytes obtained from mouse immunized with recombinant yeast SUMO protein.
SUMO-tag Monoclonal antibody specifically detects SUMO-tag in Tag samples. It is validated for ELISA, Western BlotSpecifications
SUMO-tag | |
Monoclonal | |
Unconjugated | |
SUMO; SUMO tag | |
Purified recombinant yeast SUMO protein | |
40 μg | |
Primary | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
4G11E9 | |
PBS with 0.02% sodium azide; pH 7.4 | |
Mouse | |
Protein A | |
RUO | |
-20°C or -80°C if preferred | |
Lyophilized | |
Tag |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.