Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SUMO2/SUMO3/SUMO4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103276
Description
Antibody detects endogenous levels of total Sumo2/3/4.
SUMO proteins and ubiquitin post translationally modify numerous cellular proteins and affect their metabolism and function. However, unlike ubiquitination, which targets proteins for degradation, sumoylation participates in a number of cellular processes, such as nuclear transport, transcriptional regulation, apoptosis, and protein stability (Su and Li, 2002).
Specifications
SUMO2/SUMO3/SUMO4 | |
Polyclonal | |
Unconjugated | |
SUMO3 | |
2810014B19Rik; D10Ertd345e; dJ281H8.4; HSMT3; IDDM5; insulin-dependent diabetes mellitus 5; sentrin 2; sentrin-2; small ubiquitin-like modifier 2; small ubiquitin-like modifier 3; small ubiquitin-like modifier 4; small ubiquitin-like modifier 4 protein; Small ubiquitin-like protein 4; Small ubiquitin-related modifier 2; small ubiquitin-related modifier 3; Small ubiquitin-related modifier 4; SMT3 (supressor of mif two, 3) homolog 1; SMT3 homolog 1; SMT3 homolog 2; SMT3 suppressor of mif two 3 homolog 1; SMT3 suppressor of mif two 3 homolog 2; SMT3 suppressor of mif two 3 homolog 2 (yeast); SMT3 suppressor of mif two 3 homolog 2-like; SMT3 suppressor of mif two 3 homolog 3; SMT3 suppressor of mif two 3 homolog 3 (yeast); SMT3 suppressor of mif two 3 homolog 4; SMT3 supressor of mif two 3 homolog 2; SMT3A; Smt3b; SMT3H1; SMT3H2; SMT3H4; Sumo2; SUMO-2; Sumo2l; SUMO3; SUMO-3; SUMO4; SUMO-4; Ubiquitin-like protein SMT3A; ubiquitin-like protein SMT3B | |
Rabbit | |
Affinity chromatography | |
RUO | |
170930, 20610, 387082, 499417, 6612, 6613, 682787, 690244 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P55854, P61956, P61957, P61959, Q5XIF4, Q6EEV6, Q9Z172 | |
Sumo2, SUMO3, SUMO4 | |
A synthesized peptide derived from human SUMO2(Accession P61956), corresponding to amino acid residues M1-Q51. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction