Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ SUMO3 Recombinant Protein

Supplier:  Abnova™ H00006612P0110

 View more versions of this product

Catalog No. 89-010-749


Add to Cart

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH08420
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
36.96
8
Glutathione Sepharose 4 Fast Flow
10 ug
MSEEKPKEGVKTENDHINLKVAGQDGSVVQFKIKRHTPLSKLMKAYCERQGLSMRQIRFRFDGQPINETDTPAQLEMEDEDTIDVFQQQTGGVPESSLAGHSF
SMT3A/SMT3H1/SUMO-3
SUMO3
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
6612
SUMO3 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SUMO3
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit