Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SUN-SRi™ 8mm Vial Crimp Seals

Thermo Scientific caps and vials are available in a wide variety of sizes and configurations to fit the needs of many applications.

$606.50

Specifications

Closure Type Crimp Cap
Closure Material Aluminum
Thread Size 8 mm
Pre-slit No
View More Specs

Products 3
Catalog Number Mfr. No. Material (Septa) Price Quantity & Availability  
Catalog Number Mfr. No. Material (Septa) Price Quantity & Availability  
14-823-230
View Documents
Thermo Scientific™
201 876
PTFE/Silicone/PTFE
Case of 1000 for $606.50
Add to cart
 
14-823-424
View Documents
Thermo Scientific™
502 184
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
14-823-425
View Documents
Thermo Scientific™
502 192
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Specifications

Specifications

Crimp Cap
Aluminum
8 mm
No
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit