Learn More
Thermo Scientific™ SUN-SRi™ Crimp Caps
Thermo Scientific caps and vials are available in a wide variety of sizes and configurations to fit the needs of many applications.
Supplier: Thermo Scientific™ 405 043
Description
Thermo Scientific caps and vials are available in a wide variety of sizes and configurations to fit the needs of many applications.
- Tin plate caps or two-part composite tin plate and aluminum cap with or without septum
- Two-part caps feature aluminum skirts that make the closures easier to crimp and remove
- 20mm Tin Plate
Specifications
PTFE/Silicone Septum | |
Aluminum | |
Crimp Cap |
Red | |
500/Pk. | |
20 mm |