Learn More
Thermo Scientific SUN-SRi Crimp Caps
Thermo Scientific caps and vials are available in a wide variety of sizes and configurations to fit the needs of many applications.
$136.50 - $1104.00
Catalog Number | Mfr. No. | Color | Quantity | Liner Material | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Quantity | Liner Material | Price | Quantity & Availability | |||
22-030207
|
Thermo Scientific
405 039 |
Blue | 500/Pk. | PTFE/Silicone Septum |
Pack of 500 for $1,104.00
|
|
22-030213
![]() |
Thermo Scientific
502 370 |
Blue | 500/Pk. | Without Septum |
Pack of 500 for $147.70
|
|
22-030215
|
Thermo Scientific
504 370 |
Red | 1000 Case | Without Septum |
Case of 1000 for $456.00
|
|
22-030209
|
Thermo Scientific
405 043 |
Red | 500/Pk. | PTFE/Silicone Septum |
Pack of 500 for $929.00
|
|
22-030214
|
Thermo Scientific
504 368 |
Silver | 500/Pk. | Without Septum |
Pack of 500 for $136.50
|
|
Description
Thermo Scientific caps and vials are available in a wide variety of sizes and configurations to fit the needs of many applications.
- Tin plate caps or two-part composite tin plate and aluminum cap with or without septum
- Two-part caps feature aluminum skirts that make the closures easier to crimp and remove
- 20mm Tin Plate
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Crimp Cap | |
Aluminum | |
20 mm |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.