Thermo Scientific™ SUN-SRi™ Crimp Caps Available on GSA/VA Contract for Federal Government customers only.
Manufacturer: Thermo Scientific™ 405 039
 View more versions of this product
Catalog No. 22-030207
  Check Availability
Add to Cart
    Specifications
    Thread Size 20mm
    Closure Material Aluminum
    Closure Type Crimp
    Color Blue
    Quantity 500/Pack
    Type Seal
    Material Closure Aluminum
    Material (Liner) PTFE/Silicone Septum
  • Description & Specifications
Related Products
Product Content Feedback