Learn More
Thermo Scientific™ SUN-SRi™ Glass Microsampling Vials
Good for use when working with small samples or samples that must be evaporated/reconstituted without using multiple vials
Manufacturer:  Thermo Scientific™ 500 855
 View more versions of this product
Catalog No. 14-823-360
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Amber
12mm
Ideal when sample evaporation and reconstitution may be necessary without transferring samples
0.011 oz.
1.25 in.
32mm
0.01 oz.
Silica Glass
350μL
0.47 in.
Crimp
100 Pack
350μL
0.011 oz., 350μL