Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ SUN-SRi™ Luer-LOCK Syringes
Offered in a range of sizes, from 2mL to 20mL, sterile and non-sterile.
Supplier: Thermo Scientific™ 201 635
Description
- Chemically resistant, inert polypropylene and polyethylene
- No rubber plunger seals or silicone lubricants that may cause sample contamination
- Bulk packaged, non sterile-no paper wrap waste
- Luer-LOCK™ connection
Specifications
Plastic | |
Yes | |
10 mL | |
Luer Lock |
Luer-lok Syringe | |
0.5 mL | |
Centered | |
Sterile |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title