Learn More
Thermo Scientific™ SUN-SRi™ Magnetic Screw-Thread Closures
Sun-SRI provides high quality vials, caps, seals, septa, and closures to meet or exceed original manufacturer's equipment specifications and optimize efficiencies in the laboratory.
Supplier: Thermo Scientific™ 405 035
Description
- 18mm closures fit both the 10 and 20mL screw-top headspace vials
- Can be used with Gerstel™, CTC/Leap™ Technologies, and other magnetic transport autosamplers
Specifications
Chlorobutyl/PTFE | |
With PTFE/Chlorobutyl Septa | |
18 mm |