Thermo Scientific™ SUN-SRi™ Manual Crimpers Available on GSA/VA Contract for Federal Government customers only.
High-quality construction for durability and long life
Manufacturer: Thermo Scientific™ 200 974
 View more versions of this product
Catalog No. 14-823-504
  Check Availability
Add to Cart
    Specifications
    For Use With Standard aluminum crimp seal
    Type 200mm Pliers-Type Decapper
    For Use With (Application) Attach and remove seals from 20mm crimp top vials
  • Description & Specifications
Related Products
Product Content Feedback