Learn More
Thermo Scientific™ SUN-SRi™ StepVial System™ Kits Available on GSA/VA Contract for Federal Government customers only.
StepVial w/standard graduated I-D™ patch and your choice of preassembled seals with septa
Manufacturer:  Thermo Scientific™ 500 726
 View more versions of this product
Catalog No. 14-823-352
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Seal w/PTFE/Silicone
Clear I-D Borosilicate Glass
Snap/Crimp
100 Pack
1.25 in.
2.0mL
2mL
Orange
32mm
12mm
Yes
Accessories