Thermo Scientific™ SUN-SRi™ Titan 30mm Syringe Filters Available on GSA/VA Contract for Federal Government customers only.
Banded polypropylene housing ensures lab safety preventing the housing from bursting
Manufacturer: Thermo Scientific™ 41225NN
 View more versions of this product
Catalog No. 14-823-283
  Check Availability
Add to Cart
    Specifications
    Color Code Yellow
    Housing Material Banded and Color-coded Polypropylene
    Volume (Metric) 2 to 100mL
    Pore Size 1.5μm
    Color Yellow
    Type Syringe Filter
    Quantity 100/Pk.
    Diameter (Metric) 30mm
    Material Nylon (NYL)
  • Description & Specifications
Related Products
Product Content Feedback