Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sunflower Seed Oil, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
20 L
Packaging:
Amber Glass Bottle
Steel Can
8001-21-6
8001-21-6

S1929, 8001-21-6

Spectrum™ Sunflower Seed Oil is, as its name suggests, the oil of sunflower seeds that is extracted by the compression of the seeds. This non-volatile oil is most commonly used in food and as a frying oil. It is also used in cosmetics as an emollient. It is also mixed with diesel to run diesel combustion engines.

Packaging Amber Glass Bottle
Saponification Value 188-194
Quantity 4 L
Specific Gravity 0.914-0.924
CAS Max % 1
Grade Reagent