Learn More
Selleck Chemical LLCSupplier Diversity Partner Sunitinib Malate 50mg 341031-54-7  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S104250MG
Sunitinib Malate is a multi-targeted RTK inhibitor targeting VEGFR2 (Flk-1) and PDGFRβ with IC50 of 80 nM and 2 nM in cell-free assays, and also inhibits c-Kit. Sunitinib Malate effectively inhibits autophosphorylation of Ire1α. Sunitinib Malate increases both death receptor and mitochondrial-dependent apoptosis.
Catalog No. 50-649-1