Learn More
American Educational Products Sunpower House Products for Science Education
Teach the principles of passive solar heating.
Specifications
Product Type Solar Energy Apparatus
View More Specs
Includes: Thermometer, water storage container, dark and light modular floor panels, a combination insulation/reflector panel, teacher's guide
Products 1
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
S42212B Products for Science Education
View Documents
American Educational Products
4706
Thermometer, Water storage container, dark and light modular floor panels, a combination insulation/reflector panel, teachers guide This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Part Not Found
N/A
Description
Description
A working model of a passive solar house that teaches students concepts such as the Greenhouse Effect, utilization of solar heat energy, advantages of different materials to absorb solar radiation, use of insulation and reflectors, and energy conservation.
Roof of the house may be opened to:
  • Insert the thermometer
  • Change the color of the floor panels
  • Replace the solar mass with water or rock heat storage
Order Info
Dimensions: 11 x 11 x 8 in. (27.9 x 27.9 x 20.3cm)
Specifications
Specifications
Solar Energy Apparatus