Learn More
Aat BioquestSupplier Diversity Partner SunRed™ Phosphate  DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 11629
Phosphatase-catalyzed hydrolysis of Sun Red phosphate (SRP) yields the Sun Red fluorophore that can be excited with the 633 nm laser with emission of ~660 nm.
Catalog No. 50-195-3373