Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Expedeon SunScript™ One Step RT-PCR Kit

Easy and reliable system designed for fast RT-PCR reactions using reverse transcriptase.

Specifications

Product Type SunScript™ One Step RT-PCR Kit
Concentration Enzyme mix: 1 x 45μL, 2X Reaction Buffer: 1 x 1000μL, Water: 1 x 1000μL
For Use With (Application) Reverse transcription
Format Liquid
Kit Type RT-PCR Kit
View More Specs

Products 1
Catalog Number Mfr. No. No. of Reactions Quantity Quantity Price Quantity & Availability  
Catalog Number Mfr. No. No. of Reactions Quantity Quantity Price Quantity & Availability  
EX430025 View Documents Expedeon
430025
25 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Allows for speed optimization of the protocol, higher specificity of the reaction
  • Can be completed in as little as 5 minutes
  • Contains SunScript™ Reverse Transcriptase RNaseH+, a high quality HotStart Taq Polymerase, RNase inhibitor, and 2X buffer containing dNTPs, salts, stabilizers and the double stranded DNA-binding dye
  • Performs the reverse transcription step at temperatures up to 75°C
Specifications

Specifications

SunScript™ One Step RT-PCR Kit
Reverse transcription
RT-PCR Kit
Enzyme mix: 1 x 45μL, 2X Reaction Buffer: 1 x 1000μL, Water: 1 x 1000μL
Liquid
Store at -20°C
Safety and Handling

Safety and Handling

ShelfLife : One year

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit