Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Expedeon SunScript™ Reverse Transcriptase RNaseH+

Consists of RNA and DNA-dependent polymerase activity as well as RNaseH activity.

Specifications

Product Type SunScript™ Reverse Transcriptase RNaseH+
RNase Reverse Transcriptase RNaseH+
For Use With (Application) Reverse transcription
Includes SunScript RT RNaseH+ (1 x 15μL), 5X Reaction Buffer (1 x 40μL), DTT (1 x 20μL)
Kit Type Reverse transcriptase kit
View More Specs

Products 3
Catalog Number Mfr. No. Concentration No. of Reactions Price Quantity & Availability  
Catalog Number Mfr. No. Concentration No. of Reactions Price Quantity & Availability  
EX412050 View Documents Expedeon
412050
RT RNaseH+: 1 x 75μL, 5X Reaction Buffer: 1 x 200μL, DTT: 1 x 100μL 50 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
EX411010 View Documents Expedeon
411010
RT RNaseH+: 1 x 15μL, 5X Reaction Buffer: 1 x 40μL, DTT: 1 x 20μL 10 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
EX413200 View Documents Expedeon
413200
RT RNaseH+: 1 x 300μL, 5X Reaction Buffer: 1 x 800μL, DTT: 1 x 400μL 200 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Active up to temperatures of 85°C
  • Engineered for high thermostability based on the well characterized HIV-1 RT
  • Synthesizes cDNA of up to 11 kbp in length in only 5 minutes
  • Free of endonucleases, exonucleases and ribonucleases
  • Includes 5X Reaction buffer, DTT
Specifications

Specifications

SunScript™ Reverse Transcriptase RNaseH+
Reverse transcription
Reverse transcriptase kit
-20°C
Reverse Transcriptase RNaseH+
SunScript RT RNaseH+ (1 x 15μL), 5X Reaction Buffer (1 x 40μL), DTT (1 x 20μL)
More than 95%
Safety and Handling

Safety and Handling

ShelfLife : One year

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit