Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUOX Mouse anti-Human, Clone: OTI1A12, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526763

Catalog No. PIMA526763


Add to cart

Description

Description

Sulfite oxidase is a homodimeric protein localized to the intermembrane space of mitochondria. Each subunit contains a heme domain and a molybdopterin-binding domain. The enzyme catalyzes the oxidation of sulfite to sulfate, the final reaction in the oxidative degradation of the sulfur amino acids cysteine and methionine. Sulfite oxidase deficiency results in neurological abnormalities which are often fatal at an early age. Alternative splicing results in multiple transcript variants encoding identical proteins.
Specifications

Specifications

SUOX
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P51687
sulfite oxidase, mitochondrial
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6821
Immunohistochemistry (Paraffin), Western Blot
OTI1A12
Unconjugated
SUOX
Liquid
SUOX
Human recombit protein fragment corresponding to amino acids 320-545 of SUOX produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit